Human recombinant Nuclear TF Y subunit alpha (from E. coli)
Fournisseur: ProSci
PRSI92-233EA 761 EUR InStock
PRSI92-233
Human recombinant Nuclear TF Y subunit alpha (from E. coli)
Protéines et Peptides
Nuclear Transcription Factor Y Subunit alpha (NFYA) is a member of the NFYA/HAP2 subunit family. NFYA founctions as a heterotrimeric transcription factor , which is composed of three components, NF-YA, NF-YB and NF-YC, binds to CCAAT motifs in the promoter regions in a variety of genes. NFYA forms a highly conserved transcription factor which stimulates the transcription of various genes by recognizing and binding to a CCAAT motif in promoters, for example in type 1 collagen, albumin and beta-actin genes.
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité