Human Recombinant Hepcidin (from E. coli)
Fournisseur: ProSci
PRSI92-234EA 789 EUR InStock
PRSI92-234
Human Recombinant Hepcidin (from E. coli)
Protéines et Peptides
Hepcidin(HAMP)is a secreted protein that belongs to the hepcidin family.It is expressed in liver, heart and brain. It is involved in the maintenance of iron homeostasis, and it is necessary for the regulation of iron storage in macrophages, and for intestinal iron absorption. The preproprotein is post-translationally cleaved into mature peptides of 20, 22 and 25 amino acids, and these active peptides are rich in cysteines, which form intramolecular bonds that stabilize their beta sheet structures.
Fusion-Tag: N-GST tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité