Human recombinant MIF (from E. coli)
Fournisseur: ProSci
PRSI92-250EA 789 EUR InStock
PRSI92-250
Human recombinant MIF (from E. coli)
Protéines et Peptides
Human MIF is a 12.5 kDa, 115 amino acid (aa) nonglycosylated polypeptide that is synthesised without asignal sequence .Secretion occurs nonclassically via an ABCA1 transporter.Pro-inflammatory cytokine.Involved in the innate immune response to bacterial pathogens. The expression of MIF at sites ofinflammation suggests a role as mediator in regulating the function of acrophages in host defense.Counteracts the anti-inflammatory activity of glucocorticoids. Has phenylpyruvate tautomerase anddopachrome tautomerase activity (in vitro), but the physiological substrate is not known. It is not clearwhether the tautomerase activity has any physiological relevance, and whether it is important for cytokineactivity.
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité