Human recombinant Fibronectin fragment (from E. coli)
Fournisseur: ProSci
PRSI92-255EA 306 EUR InStock
PRSI92-255
Human recombinant Fibronectin fragment (from E. coli)
Protéines et Peptides
Fibronectin1(FN1) is a secreted protein and contains 12 fibronectin type-I domains,fibronectin type-II domains and 16 fibronectin type-III domains.Recombinant human fibronectin fragment, is a protein of ~63 kDa containing a central cell-binding domain, a high affinity heparin-binding domain II,and CS1 site within the alternatively spliced III CS region of human fibronectin. Cells bind to a VLA-4 ligand, a CS-I site, and a VLA-5 ligand, a cell attachment domain, and virus vectors binds to a heparin binding domain II, which co-locates the cell and the virus vector on NovoNectin. This process enhances the density of both cells and vectors, and facilitates the gene transduction in the result.
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité