Human recombinant Neuritin (from E. coli)
Fournisseur: ProSci
PRSI92-259EA 525 EUR InStock
PRSI92-259
Human recombinant Neuritin (from E. coli)
Protéines et Peptides
Neuritin/NRN1 is a member of the neuritin family and can be expressed in postmitotic-differentiating neurons of the developmental nervous system and neuronal structures associated with plasticity in the adult. Neuritin/NRN1 promotes neurite outgrowt, arborisation and neuritogenesis. The protein contains a consensus cleavage signal found in glycosylphoshatidylinositol (GPI)-anchored proteins.Overexpression of the encoded protein may be associated with astrocytoma progression.
Fusion-Tag: N-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité