Human recombinant Polyubiquitin-B (from E. coli)
Fournisseur: ProSci
PRSI92-271EA 584 EUR InStock
PRSI92-271
Human recombinant Polyubiquitin-B (from E. coli)
Protéines et Peptides
Polyubiquitin-B(UBB) is one of Ubiquitins. Ubiquitin is one of the most conserved proteins known in eukaryotic organisms. Ubiquitin is required for ATP-dependent, non-lysosomal intracellular protein degradation of abnormal proteins and normal proteins with a rapid turnover. Ubiquitin is covalently bound to proteins to be degraded, and presumably labels these proteins for degradation.
Fusion-Tag: N-GST, 6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité