Human recombinant UQCRH (from E. coli)
Fournisseur: ProSci
PRSI92-278EA 761 EUR InStock
PRSI92-278
Human recombinant UQCRH (from E. coli)
Protéines et Peptides
Cytochrome b-c1 complex subunit 6, mitochondrial (UQCRH) belongs to the UQCRH/QCR6 family, it is a subunit of the respiratory chain protein Ubiquinol Cytochrome c Reductase. This is a component of the ubiquinol-cytochrome c reductase complex (complex III or cytochrome b-c1 complex), which is part of the mitochondrial respiratory chain. UQCRH may mediate formation of the complex between cytochromes c and c1. It may mediate formation of the complex between cytochromes c and c1.
Fusion-Tag: N-GST tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité