Human recombinant protachykinin-1 (from E. coli)
Fournisseur: ProSci
PRSI92-287EA 789 EUR InStock
PRSI92-287
Human recombinant protachykinin-1 (from E. coli)
Protéines et Peptides
Protachykinin-1(TAC1) is a secreted protein and belongs to the tachykinin family. TAC1 is encoded by the TAC1 gene. This gene encodes four products of the tachykinin peptide hormone family, substance P and neurokinin A, as well as the related peptides, neuropeptide K and neuropeptide gamma. These hormones are thought to function as neurotransmitters which interact with nerve receptors and smooth muscle cells. They are known to induce behavioral responses and function as vasodilators and secretagogues.
Fusion-Tag: N-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité