Human recombinant Apolipoprotein E (from cells)
Fournisseur: ProSci
PRSI92-312EA 306 EUR InStock
PRSI92-312
Human recombinant Apolipoprotein E (from cells)
Protéines et Peptides
ApoE, a glycoprotein, is a structural component of very low density lipoprotein (vLDL) Synthesised by the liver and intestinally Synthesised chylomicrons . ApoE is also a constituent of a subclass of high density of lipoproteins (HDL) involved in cholesterol transport. ApoE mediates high affinity binding of chylomicrons and vLDL particles to the LDL receptor, allowing for specific uptake of these particles by the liver, preventing the accumulation of cholesterol rich particles in the plasma. Apolipoprotein E combines with fats (lipids) in the body to form molecules called lipoproteins and Apolipoprotein E is a major component of a specific type of lipoprotein called very low-density lipoproteins (VLDLs).
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité