Human recombinant neuritin 1-like (from cells)
Fournisseur: ProSci
PRSI92-348EA 525 EUR InStock
PRSI92-348
Human recombinant neuritin 1-like (from cells)
Protéines et Peptides
Neuritin-like protein belongs to the neuritin family. Neuritin is a GPI-anchored protein that promotes neurite outgrowth and branching of neuritic processes in primary hippocampal and cortical cells. Neuritin expression also enhances the development of motor neuron axon arbors by promoting neuromuscular synaptogenesis and by stimulating the addition of new axon branches. Neuritin is induced by neuronal activity and by the neurotrophins, BDNF and NT3. NRN1L contains a consensus cleavage signal found in glycosylphoshatidylinositol (GPI)-anchored proteins.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité