Human recombinant insulin-like 3 (from cells)
Fournisseur: ProSci
PRSI92-352EA 525 EUR InStock
PRSI92-352
Human recombinant insulin-like 3 (from cells)
Protéines et Peptides
Insulin-like 3 is a protein that in humans is encoded by the INSL3 gene. It is a secreted protein that belongs to the insulin family. It is expressed in prenatal and postnatal Leydig cells and found as well in the corpus luteum, trophoblast, fetal membranes and breast. It may act as a hormone to regulate growth and differentiation of gubernaculum, and thus mediating intra-abdominal testicular descent.It is a ligand for LGR8 receptor.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité