Human recombinant CD99 antigen-like 2 (from cells)
Fournisseur: ProSci
PRSI92-380EA 525 EUR InStock
PRSI92-380
Human recombinant CD99 antigen-like 2 (from cells)
Protéines et Peptides
CD99 Antigen-Like Protein 2 (CD99L2) belongs to the CD99 family. CD99L2 is a single-pass type I membrane protein and expressed in many tissues, with low expression in thymus. CD99L2 plays a role in a late step of leukocyte extravasation helping cells to overcome the endothelial basement membrane. CD99L2 and CD99 are involved in trans-endothelial migration of neutrophils in vitro and in the recruitment of neutrophils into inflamed peritoneum. A similar protein in mouse functions as an adhesion molecule during leukocyte extravasation. Alternate splicing results in multiple transcript variants.
Fusion-Tag: C-Fc tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité