Human recombinant Secretogranin-3 (from cells)
Fournisseur: ProSci
PRSI92-383EA 365 EUR InStock
PRSI92-383
Human recombinant Secretogranin-3 (from cells)
Protéines et Peptides
Secretogranin-3(SCG3) is a member of the chromogranin/secretogranin family of neuroendocrine secretory proteins. SCG3 is expressed in brain, heart, kidney, liver and skeletal muscle. It may serve as precursors for biologically active peptides. Some granins have been shown to function as helper proteins in sorting and proteolytic processing of prohormones; however, the function of this protein is unknown.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité