Human recombinant LMAN2L (from cells)
Fournisseur: ProSci
PRSI92-384EA 584 EUR InStock
PRSI92-384
Human recombinant LMAN2L (from cells)
Protéines et Peptides
VIP36-like protein (LMAN2L) is a single-pass type I membrane protein and contains 1 L-type lectin-like domain. It is highly expressed in skeletal muscle and kidney, and its intermediate expression levels in heart, liver and placenta, low levels in brain, thymus, spleen, small intestine and lung. LMAN2L may be involved in the regulation of export from the endoplasmic reticulum of a subset of glycoproteins. It also may function as a regulator of ERGIC-53.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité