Human recombinant MFAP4 (from cells)
Fournisseur: ProSci
PRSI92-386EA 365 EUR InStock
PRSI92-386
Human recombinant MFAP4 (from cells)
Protéines et Peptides
Microfibril-associated glycoprotein 4(MFAP4) is a secreted protein and contains 1 fibrinogen C-terminal domain, similarity to a bovine microfibril-associated protein. The protein has binding specificities for both collagen and carbohydrate. It is thought to be an extracellular matrix protein which is involved in calcium-dependent cell adhesion or intercellular interactions. The gene is located within the Smith-Magenis syndrome region.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité