Human recombinant Amiloride-binding 1 (from cells)
Fournisseur: ProSci
PRSI92-393EA 365 EUR InStock
PRSI92-393
Human recombinant Amiloride-binding 1 (from cells)
Protéines et Peptides
Amiloride-sensitive amine oxidase (AOC1) belongs to the copper/topaquinone oxidase family. The protein exists as homodimer by disulfide and mainly located in placenta and kidney. AOC1 catalyses the degradation of compounds such as putrescine, histamine, spermine, and spermidine, substances involved in allergic and immune responses, cell proliferation, tissue differentiation, tumour formation, and possibly apoptosis. Placental DAO is thought to play a role in the regulation of the female reproductive function. The activity of this protein can be inhibited by amiloride in a competitive manner. It is inhibited by amiloride, a diuretic that acts by closing epithelial sodium ion channels.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité