Human recombinant serpin B1 (from cells)
Fournisseur: ProSci
PRSI92-403EA 584 EUR InStock
PRSI92-403
Human recombinant serpin B1 (from cells)
Protéines et Peptides
SERPINB1 is a member of the serpin family and Ov-serpin subfamily. As protease inhibitors, serpins have an array of functions including regulating blood coagulation, fibrinolysis, the complement pathway, angiogenesis, inflammation, tumour suppression, extracellular matrix remodelling, and cell motility. SERPINB1 regulates the activity of the neutrophil proteases elastase, cathepsin G, proteinase-3, chymase, chymotrypsin, and kallikrein-3. Reactive bond 1 of SerpinB1 is specific for reaction with chymotrypsin-like protease such as cathepsin G, chymotrypsin or chymase. Reactive bond 2 of SerpinB1 is specific for reaction with elastase-like protease such as neutrophyl elastase, proteinase-3, pancreatic elastase or PSA. In addition, SERPINB1 also functions as a potent intracellular inhibitor of granzyme H.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité