Mouse recombinant SEMA4D
Fournisseur: ProSci
PRSI92-410EA 525 EUR InStock
PRSI92-410
Mouse recombinant SEMA4D
Protéines et Peptides
SEMA4D is a member of the semaphorin family,contains one Ig-like C2-type domain, one PSI domain and one Sema domain. SEMA4D is strongly expressed in lymphoid tissues, especially in the thymus, as well as in the nervous tissues. However, SEMA4D is expressed at lower levels in testes, brain, kidney, small intestine, prostate, heart, placenta, lung and pancreas, but not in colon and liver. SEMA4D is a cell surface receptor for PLXN1B and PLXNB2 that plays an important role in cell-cell signalling. SEMA4D is involved in a number of fundamental biological processes such as promoting reorganisation of the actin cytoskeleton, the migration of cerebellar granule cells and of endothelial cells and signalling via SRC and PTK2B/PYK2, which then mediates activation of phosphatidylinositol 3-kinase and of the AKT1 signalling cascade. Not only these, it plays a role in axonal growth cone guidance in the developing central nervous system. Semaphorin-4D / SEMA4D may play a functional role in the immune system, as well as in the nervous system. It could induce B-cells to aggregate and improves their viability (in vitro). SEMA4D is involved in regulating dendrite and axon branching and morphogenesis and promoting interaction with PLXNB1 mediates activation of RHOA.
Fusion-Tag: C-Fc tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité