Human recombinant Prion-like doppel (from cells)
Fournisseur: ProSci
PRSI92-424EA 584 EUR InStock
PRSI92-424
Human recombinant Prion-like doppel (from cells)
Protéines et Peptides
Prion-like protein doppel is a 152 amino acids protein that belongs to the prion family. The protein encoded by this gene is found on chromosome 20, approximately 20 kbp downstream of the gene encoding cellular prion protein, to which it is biochemically and structurally similar. It is a membrane glycosylphosphatidylinositol-anchored glycoprotein that is found predominantly in testis. Mutations in this gene may lead to neurological disorders.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité