Human recombinant Contactin-2 (from cells)
Fournisseur: ProSci
PRSI92-426EA 584 EUR InStock
PRSI92-426
Human recombinant Contactin-2 (from cells)
Protéines et Peptides
Contactin-2 (CNTN2) is encoded by the CNTN2 gene,which belongs to the immunoglobulin superfamily and contactin family. It contains 4 fibronectin type-III domains and 6 Ig-like C2-type domains. It is a glycosylphosphatidylinositol (GPI)-anchored neuronal membrane protein that functions as a cell adhesion molecule. CNTN2 may play a role in the formation of axon connections in the developing nervous system. In conjunction with another transmembrane protein, CNTNAP2, contributes to the organisation of axonal domains at nodes of Ranvier by maintaining voltage-gated potassium channels at the juxtaparanodal region. It may also be involved in glial tumourigenesis and may provide a potential target for therapeutic intervention.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité