Human recombinant Kell (from cells)
Fournisseur: ProSci
PRSI92-428EA 584 EUR InStock
PRSI92-428
Human recombinant Kell (from cells)
Protéines et Peptides
Kell blood group glycoprotein (KEL) is a single-pass type II membrane protein which belongs to the peptidase M13 family. It is expressed in Expressed at high levels in erythrocytes and testis, and, at lower levels, in skeletal muscle, tonsils, lymph node, spleen and appendix. KEL has been shown zinc endopeptidase with endothelin-3-converting enzyme activity. It cleaves EDN1, EDN2 and EDN3, with a marked preference for EDN3. It links via a single disulfide bond to the XK membrane protein that carries the Kx antigen.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité