Human Recombinant HCLS1 (from Cells)
Fournisseur: ProSci
PRSI92-435EA 789 EUR InStock
PRSI92-435
Human Recombinant HCLS1 (from Cells)
Protéines et Peptides
Hematopoietic lineage cell-specific protein (HCLS1) is a protein that in humans is encoded by the HCLS1 gene. It is expressed only in tissues and cells of hematopoietic origin. It is substrate of the antigen receptor-coupled tyrosine kinase and plays a role in antigen receptor signalling for both clonal expansion and deletion in lymphoid cells. It may also be involved in the regulation of gene expression.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité