Human Recombinant 4-1BB (from Cells)
Fournisseur: ProSci
PRSI92-439EA 388 EUR InStock
PRSI92-439
Human Recombinant 4-1BB (from Cells)
Protéines et Peptides
tumour necrosis factor receptor superfamily member 9(TNFRSF9) is an inducible T cell surface protein belonging to the TNF receptor superfamily. It is a single-pass type I membrane protein which contains 4 TNFR-Cys repeats. The human and mouse proteins share 60% amino acid sequence identity. It is absent from naive T cells, but upregulated and continually expressed following T cell activation. It is a receptor for TNFSF9/4-1BBL, and possibly active during T cell activation.
Fusion-Tag: C-Fc-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité