Human Recombinant GDF8 (from Cells)
Fournisseur: ProSci
PRSI92-443EA 1060 EUR InStock
PRSI92-443
Human Recombinant GDF8 (from Cells)
Protéines et Peptides
Growth/differentiation factor 8(Mstn, GDF-8) is a member of the bone morphogenetic protein (BMP) family and the TGF-beta superfamily. This group of proteins is characterised by a polybasic proteolytic processing site which is cleaved to produce a mature protein containing seven conserved cysteine residues. It is expressed specifically in developing and adult skeletal muscle. It exists as a homodimer, and interacts with WFIKKN2, leading to inhibit its activity. This protein can act specifically as a negative regulator of skeletal muscle growth. It regulates cell growth and differentiation in both embryonic and adult tissues.
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité