Mouse recombinant TYRO3
Fournisseur: ProSci
PRSI92-447EA 306 EUR InStock
PRSI92-447
Mouse recombinant TYRO3
Protéines et Peptides
Dtk, also called Tyro3, belongs to the TAM receptor family of receptor protein tyrosine kinases (RPTKs) composed of three receptors Tyro3, Axl, and Mer. These receptors share a characteristic molecular structure of two immunoglobulin-like and two fibronectin type III repeats and have been best characterised for their roles in immune regulation, fertility, thrombosis and phagocytosis. Gas6 and protein S have been identified as ligands for these receptors. Gas6 binding induces tyrosine phosphorylation and downstream signalling pathways that can lead to cell proliferation, migration, or the prevention of apoptosis. Tyro3 and Axl play important regulatory roles in a variety of tissues, including the central nervous, reproductive, immune, and vascular systems. Tyro3 is widely expressed during embryonic development and preferentially expressed during neurogenesis in the central nervous system.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-mFc tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité