Mouse recombinant TACI
Fournisseur: ProSci
PRSI92-452EA 525 EUR InStock
PRSI92-452
Mouse recombinant TACI
Protéines et Peptides
tumour necrosis factor receptor superfamily member 13B, also known as TNFRSF13B or more commonly as TACI (transmembrane activator and CAML interactor), is a transmembrane receptor protein found predominantly on the surface of B cells, which are an important part of the immune system.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-Fc tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité