Human recombinant PTH1 receptor (from cells)
Fournisseur: ProSci
PRSI92-453EA 525 EUR InStock
PRSI92-453
Human recombinant PTH1 receptor (from cells)
Protéines et Peptides
Parathyroid hormone/parathyroid hormone-related peptide receptor also known as parathyroid hormone 1 receptor (PTH1R) is a protein that in humans is encoded by the PTH1R gene, Belongs to the G-protein coupled receptor 2 family.PTH1R functions as a receptor for parathyroid hormone (PTH) and for parathyroid hormone-related protein (PTHrP), also called parathyroid hormone-like hormone (PTHLH).
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité