Human recombinant VSTM1 (from cells)
Fournisseur: ProSci
PRSI92-469EA 584 EUR InStock
PRSI92-469
Human recombinant VSTM1 (from cells)
Protéines et Peptides
V-set and transmembrane domain-containing protein 1 is a single-pass membrane protein. VSTM1 Contains 1 Ig-like V-type domain, in humans is encoded by the VSTM1 gene. It expressed on myeloid (neutrophils, eosinophils and monocytes) but not on lymphoid cells. It behaves as a cytokine, promoting IL17A secretion by CD4+ T-cells, and differentiation and activation of IL17 producing helper T-cells (TH17). Inhibitory immune receptor involved in the regulation of phagocytes.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité