Human Recombinant Hepatocyte Growth Factor (from Cells)
Fournisseur: ProSci
PRSI92-470EA 584 EUR InStock
PRSI92-470
Human Recombinant Hepatocyte Growth Factor (from Cells)
Protéines et Peptides
Hepatocyte growth factor/scatter factor (HGF/SF) is a paracrine cellular growth, motility and morphogenic factor. It belongs to the peptidase S1 family and Plasminogen subfamily, contains 4 kringle domains, 1 PAN domain and 1 peptidase S1 domain. HGF regulates cell growth, cell motility, and morphogenesis by activating a tyrosine kinase signalling cascade after binding to the proto-oncogenic c-Met receptor. HGF is secreted by mesenchymal cells and acts as a multi-functional cytokine on cells of mainly epithelial origin. Its ability to stimulate mitogenesis, cell motility, and matrix invasion gives it a central role in angiogenesis, tumourogenesis, and tissue regeneration.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité