Human recombinant TGF beta 2 (from cells)
Fournisseur: ProSci
PRSI92-477EA 761 EUR InStock
PRSI92-477
Human recombinant TGF beta 2 (from cells)
Protéines et Peptides
Transforming growth factor beta-2 (TGF- beta 2) is a secreted protein which belongs to the TGF-beta family. It is known as a cytokine that performs many cellular functions and has a vital role during embryonic development. The precursor is cleaved into mature TGF-beta-2 and LAP, which remains non-covalently linked to mature TGF-beta-2 rendering it inactive. It is an extracellular glycosylated protein. It is known to suppress the effects of interleukin dependent T-cell tumours. Defects in TGFB2 may be a cause of non-syndromic aortic disease (NSAD).
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité