Mouse recombinant CXCL15
Fournisseur: ProSci
PRSI92-483EA 761 EUR InStock
PRSI92-483
Mouse recombinant CXCL15
Protéines et Peptides
Mouse C-X-C motif chemokine 15, also known as Cxcl15, is a secreted protein which is member of the ELR motif-containing CXC chemokines. It expressed at low levels in fetal lung, the expression restricted to the lung, produced by bronchoepithelial cells and is released into the airways. It plays an important role in lung-specific neutrophil trafficking during normal and inflammatory conditions. It also appears chemotactic for neutrophils.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité