Human Recombinant ECH1 (from E. coli)
Fournisseur: ProSci
PRSI92-505EA 584 EUR InStock
PRSI92-505
Human Recombinant ECH1 (from E. coli)
Protéines et Peptides
Human delta(3,5)-Delta(2,4)-dienoyl-CoA isomerase(ECH1) is a member of the hydratase/isomerase superfamily and contains a C-terminal peroxisomal targeting sequence and localises to peroxisomes. ECH1 shows high sequence similarity to enoyl-CoA hydratases of several species, particularly within a conserved domain characteristic of these proteins. The rat orthologlocalises to the matrix of both the peroxisome and mitochondria.It can isomerize 3-trans, 5-cis-dienoyl-CoA to 2-trans,4-trans-dienoyl-CoA, indicating that it is a delta3,5-delta2,4-dienoyl-CoA isomerase. ECH1 plays an important role in the auxiliary step of the fatty acid beta-oxidation pathway.
Fusion-Tag: N-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité