Mouse recombinant TGF beta 2
Fournisseur: ProSci
PRSI92-531EA 761 EUR InStock
PRSI92-531
Mouse recombinant TGF beta 2
Protéines et Peptides
Transforming growth factor beta 2 (TGF- beta 2) is a member of TGF-beta superfamily that shares a characteristic cysteine knot structure. Mice with TGF- beta 2 gene deletion show defects in development of cardiac, lung, craniofacial, limb, spinal column, eye, inner ear and urogenital systems. All TGF- beta isoforms signal via the same heteromeric receptor complex, consisting of a ligand binding TGF- beta receptor type II (T beta R-II), and a TGF- beta receptor type I (T beta R-I). Signal transduction from the receptor to the nucleus is mediated via SMADs. TGF- beta expression is found in cartilage, bone, teeth, muscle, heart, blood vessels, haematopoitic cells, lung, kidney, gut, liver, eye, ear, skin, and the nervous system.
  • This recombinant protein can be used for biological assays. For research use only.
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité