Mouse recombinant kallikrein 7
Fournisseur: ProSci
PRSI92-580EA 761 EUR InStock
PRSI92-580
Mouse recombinant kallikrein 7
Protéines et Peptides
Kallikrein7, also named as stratum corneum chymotryptic enzyme (SCCE), is a secreted protein of the Kallikrein-related peptidase (KLK) family. This family contains fifteen homologous secreted serine endopeptidases and plays a significant role in various physiological processes, including skin desquamation, semen liquefaction, neural plasticity, and body fluid homeostasis. In skin KLK5, KLK 7 and KLK14 are able to degrade corneodesmosomes, which leads to desquamation of skin surface cells. KLK activation is believed to be mediated through highly organised proteolytic cascades, regulated through a series of feedback loops, inhibitors, auto-degradation and internal cleavages. Studies have shown that one potential physiological activator for KLK7 is KLK5. Along with KLK14, these three kallikreins form a proteolytic cascade in the stratum corneum. KLK7 is primarily expressed in the skin but is also detected at relatively high levels in oesophagus, heart, liver, central nervous system, kidney, pancreas, mammary and salivary glands.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité