Mouse recombinant Exostosin-like 2
Fournisseur: ProSci
PRSI92-582EA 584 EUR InStock
PRSI92-582
Mouse recombinant Exostosin-like 2
Protéines et Peptides
Exostosin-like 2 (EXTL2) is a member of the exostosin (EXT)-related family which contains five members: EXT1, EXT2, EXTL1, EXTL2, and EXTL3. Studies have shown that EXT gene family members have the activities of heparan sulphate-synthesising glycosyltransferases. EXT1 and EXT2, which have been identified as causal genes for hereditary multiple exostoses, have HS-GlcAT-II and GlcNAcT-II activities. EXTL1 has GlcNAcT-II activity and EXTL3 has GlcNAcT-I and -II activities. EXTL2 has GlcNAcT-I and N-acetylgalactosaminyltransferase activities, and transfers a GlcNAc residue to the tetrasaccharide linkage region when this region is phosphorylated by a xylose kinase 1 (FAM20B) and thereby terminate chain elongation. In mice, lack of EXTL2 causes glycosaminoglycan (GAG) overproduction and structural changes of GAGs associated with pathological processes.
Fusion-Tag: N-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité