Mouse recombinant GH Receptor
Fournisseur: ProSci
PRSI92-595EA 412 EUR InStock
PRSI92-595
Mouse recombinant GH Receptor
Protéines et Peptides
Growth hormone receptor is a transmembrane receptor for growth hormone (GH). GH is a single-chain polypeptide that is mainly synthesised and released from the anterior pituitary gland and plays essential roles in growth, development and metabolism. GH exerts its physiological actions via GH binding to its receptor in its extracellular domain. Binding of growth hormone to the receptor leads to receptor dimerisation and the activation of an intra- and intercellular signal transduction pathway leading to growth. Growth hormone receptor has been shown to interact with SGTA, PTPN11, Janus kinase 2, Suppressor of cytokine signaling 1 and CISH.
Fusion-Tag: C-Fc tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité