Vaccinia virus recombinant B18R
Fournisseur: ProSci
PRSI92-624EA 761 EUR InStock
PRSI92-624
Vaccinia virus recombinant B18R
Protéines et Peptides
B18R, also known as B19R in the Copenhagen strain of Vaccinia, is a type I interferon (IFN)-binding protein encoded by the B18R open reading frame in the Western Reserve strain of vaccinia virus. B18R exists in two forms, soluble and membrane-bound form and has a broad species specificity. B18R has high affinity for human IFN-alpha and binds rabbit, bovine, rat, pig, and mouse IFN-alpha and IFN-beta. Secreted B18R binds to uninfected and infected cells. B18R presents at the cell surface and protects cells from the antiviral state. It has shown that binding of soluble recombinant B18R protects cultured cells from IFN and allows vaccinia virus replication.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité