Mouse recombinant limitin
Fournisseur: ProSci
PRSI92-638EA 761 EUR InStock
PRSI92-638
Mouse recombinant limitin
Protéines et Peptides
Limitin, also called IFN-ζ, is a secreted interferon (IFN)-like glycoprotein. Limitin has approximately 30% sequence homology with IFN- alpha, IFN- beta , and IFN-ω and binds to the IFN- alpha/ beta receptors. Like IFN- alpha and IFN- beta , limitin has antiproliferative, immunomodulatory, and antiviral properties, it is unique in lacking influence on myeloid and erythroid progenitors. Similar dose requirement between limitin and IFN- alpha was observed for the enhancement of cytotoxic T lymphocyte activity, the augmentation of MHC class I expression, and the growth inhibition of a myelomonocytic leukaemia cell line.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité