Human recombinant spondin 2 (from cells)
Fournisseur: ProSci
PRSI92-655EA 412 EUR InStock
PRSI92-655
Human recombinant spondin 2 (from cells)
Protéines et Peptides
Spondin-2, also referred to as mindin, belongs to the F-spondin family of secreted extracellular matrix proteins. Spondins are characterised by the presence of F-spondin domains 1 and 2 (FS1 and FS2) at the N-terminus and a thrombospondin-type 1 repeat (TSR1) domain at the C-terminus. Spondin-2 functions as a pattern-recognition molecule for bacterial and viral pathogens and as an integrin ligand for inflammatory cell recruitment and T cell priming. In addition to its roles in promoting neuron outgrowth and inhibiting both cancer and angiogenesis, Spondin-2 plays an important role in the initiation of the immune response and is involved in inflammatory processes.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité