Human recombinant SUMO3 (from cells)
Fournisseur: ProSci
PRSI92-665EA 365 EUR InStock
PRSI92-665
Human recombinant SUMO3 (from cells)
Protéines et Peptides
Small ubiquitin-like modifier (SUMO), also known as SUMO homologue and SMT3, is a member of the superfamily of ubiquitin-like polypeptides that become covalently attached to various intracellular target proteins as a way to alter their function, location, and/or half-life. Small ubiquitin-like modifiers include SUMO1, SUMO2, SUMO3, and SUMO4. Except for SUMO4, all other SUMOs are ubiquitously expressed, including in the brain. In human, SUMO2 and SUMO3 are two highly homologous proteins, collectively called SUMO2/3. Several studies suggest that SUMO3 are associated with pathogenesis in several neurological diseases, including Alzheimer's disease, Parkinson's disease, and cerebral ischemia/stroke.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité