Mouse Recombinant PDGF R alpha
Fournisseur: ProSci
PRSI92-686EA 525 EUR InStock
PRSI92-686
Mouse Recombinant PDGF R alpha
Protéines et Peptides
Platelet-derived growth factor receptors (PDGFR) are cell surface tyrosine kinase receptors for members of the platelet-derived growth factor (PDGF) family. The PDGF family consists of PDGF-A, -B, -C and -D, which form either homo- or heterodimers (PDGF-AA, -AB, -BB, -CC, -DD). The four PDGFs are inactive in their monomeric forms. PDGFs bind to the protein tyrosine kinase receptors PDGF receptor- alpha and - beta . These two receptor isoforms dimerise upon binding the PDGF dimer, leading to three possible receptor combinations, namely - alpha alpha, - beta beta and - alpha beta . PDGFR alpha and PDGFR beta are members of the class III RTK family. Inappropriate PDGFR alpha and PDGFR beta signalling has been linked to a number of proliferative disorders.
Fusion-Tag: C-Fc tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité