Human recombinant TNF receptor II (from cells)
Fournisseur: ProSci
PRSI92-700EA 525 EUR InStock
PRSI92-700
Human recombinant TNF receptor II (from cells)
Protéines et Peptides
Tumour necrosis factor receptor superfamily member 1B (TNFRSF1B) is a member of the tumour necrosis factor receptor superfamily. Human TNF RII contains four cysteinerich repeats in its ECD, which shares 58% and 56% amino acid sequence identity with the mouse and rat orthologs, respectively.TNF RII is expressed predominantly on cells of the hematopoietic lineage, such as T and natural killer cells, as well as on endothelial cells, microglia, astrocytes,neurons, oligodendrocytes, cardiac myocytes, thymocytes, and mesenchymal stem cells. TNF RII binds to the membrane bound forms of TNF alpha and Lymphotoxin alpha/TNF beta ;soluble TNF is thought to signal predominately through TNF RI. Soluble TNF RII is believed to inhibit TNF biological activity by binding TNF thereby preventing it from activating membrane TNF receptors.
Fusion-Tag: C-Fc tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité