Human recombinant CEACAM5 (from cells)
Fournisseur: ProSci
PRSI92-705EA 525 EUR InStock
PRSI92-705
Human recombinant CEACAM5 (from cells)
Protéines et Peptides
Carcinoembryonic antigen-related cell adhesion molecules (CEACAMs) belong to a group of mammalian immunoglobulin related glycoproteins. They play critical roles in cell–cell recognition. CEACAM5, also called CEA and CD66e, is characterized by having seven extracellular Ig domains and a glycosylphosphatidylinositol (GPI) anchor. CEACAM5 is expressed primarily by epithelial cells, and functions as a calcium-independent adhesion molecule through homophilic and heterophilic interactions with CEACAM1. Studies have shown that CEACAM5 is overexpressed in a majority of carcinomas, and its overexpression can protect tumour cells from apoptosis. It is commonly used as a cancer marker.
Fusion-Tag: C-Fc tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité