Human recombinant CD3 epsilon (from cells)
Fournisseur: ProSci
PRSI92-708EA 525 EUR InStock
PRSI92-708
Human recombinant CD3 epsilon (from cells)
Protéines et Peptides
T-Cell Surface Glycoprotein CD3 epsilon Chain (CD3 epsilon ) is a single-pass type I membrane protein. CD3 epsilon contains 1 Ig-like (immunoglobulin-like) domain and 1 ITAM domain. CD3 epsilon is a polypeptide encoded by the CD3E gene on chromosome 11 in humans. The T cell receptor-CD3 complex (TCR/CD3 complex) is involved in T-cell development and several intracellular signal-transduction pathways. This complex is critical for T-cell development and function, and represents one of the most complex transmembrane receptors. The T cell receptor-CD3 complex is unique in having ten cytoplasmic immunoreceptor tyrosine-based activation motifs (ITAMs). TCR/CD3 complex plays an important role in coupling antigen recognition to several intracellular signal-transduction pathways.
Fusion-Tag: C-Fc tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité