Mouse recombinant NUCB2 (from E. coli)
Fournisseur: ProSci
PRSI90-073EA 655 EUR InStock
PRSI90-073
Mouse recombinant NUCB2 (from E. coli)
Protéines et Peptides
Nucelobindin-2 (NUCB2; Nesfatin-1) was first isolated in adipocytes, but is also expressed by gastric mucosa and pancreatic beta cells. This pattern of nucleobindin-2 expression, as well as the presence of nucelobindin-2 within the plasma of rodents and humans, strongly suggests that this compound may act as a circulating regulatory factor. Nucelobindin-2 is reported to reduce food intake after administration. It is responsible for regulating appetite and production of body fat. Excess nucelobindin-2 in the brain leads to a loss of appetite, lack of nesfatin-1 in the brain leads to an increase of appetite.
Fusion-Tag: His Tag
This recombinant proteins is for research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité