Human recombinant JAG1 (from HEK293 cells)
Fournisseur: ProSci
PRSI90-084EA 400 EUR InStock
PRSI90-084
Human recombinant JAG1 (from HEK293 cells)
Protéines et Peptides
Jagged-1 is a ligand for multiple Notch receptors and involved in the mediation of Notch signalling. It is involved in cell-fate decisions during haematopoiesis. Is involved in early and late stages of mammalian cardiovascular development. Inhibits myoblast differentiation. Enhances fibroblast growth factor-induced angiogenesis (in vitro). Defects in Jagged-1 are the cause of Alagille syndrome type 1 (ALGS1) and tetralogy of Fallot (TOF).
Fusion-Tag: Fc Tag
This recombinant proteins is for research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité