Human Recombinant FTO (from E. coli)
Fournisseur: ProSci
PRSI90-120EA 820 EUR InStock
PRSI90-120
Human Recombinant FTO (from E. coli)
Protéines et Peptides
FTO (Fat mass-and obesity-associated gene) is the responsible gene for mouse ‘fused toes’ mutation. An association between FTO genotype and type 2 diabetes has been confirmed. The presence of the FTO rs9939609 A-allele was found to be positively correlated with other symptoms of the metabolic syndrome, including higher fasting insulin, glucose, triglycerides, and lower HDL-cholesterol.
Fusion-Tag: His Tag
This recombinant proteins is for research use only.
Commander maintenant
COMMANDE
SPÉCIFICATIONS
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité