Human recombinant IL37 (from E. coli)
Fournisseur: ProSci
PRSI90-207EA 1100 EUR InStock
PRSI90-207
Human recombinant IL37 (from E. coli)
Protéines et Peptides
IL-37 (IL-1F7; IL-1H4) belongs to the IL-1 family of proteins. It binds to the interleukin-18 receptor (IL-18R) but not to the IL-1 receptor. It plays an important role as a link between innate and adaptive immunity. IL-37 is a new player in inflammatory and immune responses, mediated by the IL-18/IL-18R axis. It was shown to inhibit tumour growth.
Fusion-Tag: His Tag
This recombinant proteins is for research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité