Human recombinant DR6 (from HEK293 cells)
Fournisseur: ProSci
PRSI90-226EA 400 EUR InStock
PRSI90-226
Human recombinant DR6 (from HEK293 cells)
Protéines et Peptides
DR6 is an orphan TNF receptor superfamily member belonging to a subgroup of receptors called death receptors. Expressed ubiquitously with high expression in lymphoid organs, heart, brain, and pancreas. Broadly expressed by developing neurons where it functions as pro-apoptotic factor. Recently, it has been reported that interaction with the beta-amyloid precursor protein (APP) activates a widespread caspase-dependent self-destruction program dependent on caspase-6.
Fusion-Tag: Fc Tag
This recombinant proteins is for research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité