Human recombinant OX40 (from HEK293 cells)
Fournisseur: ProSci
PRSI90-227EA 537 EUR InStock
PRSI90-227
Human recombinant OX40 (from HEK293 cells)
Protéines et Peptides
OX40 binds to receptors on T-cells, preventing them from dying and subsequently increasing cytokine production. OX40 has a critical role in the maintenance of an immune response. OX40:Fc was shown to prevent OX40 from reaching the T-cell receptors, thus reducing the T-cell response. Experiments in mice have demonstrated that OX40:Fc can reduce the symptoms associated with the cytokine storm (an immune overreaction) while allowing the immune system to fight off the virus successfully.
Fusion-Tag: Fc Tag
This recombinant proteins is for research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité